missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lymphotoxin beta/TNFSF3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£170.00 - £501.00
Specifications
| Antigen | Lymphotoxin beta/TNFSF3 |
|---|---|
| Clone | 135105 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728978
![]() |
R&D Systems
MAB1684-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18132511
![]() |
R&D Systems
MAB1684 |
500 μg |
£501.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lymphotoxin beta/TNFSF3 Monoclonal specifically detects Lymphotoxin beta/TNFSF3 in Human samples. It is validated for Western Blot.Specifications
| Lymphotoxin beta/TNFSF3 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| LTB, lymphotoxin beta (TNF superfamily, member 3), lymphotoxin-beta, p33, TNFCTNF-C, TNFSF3, TNFSF3LT-beta, Tumor necrosis factor C, Tumor necrosis factor ligand superfamily member 3 | |
| LTB | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 135105 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 4050 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Lymphotoxin alpha/beta | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Lymphotoxin alpha beta heterotrimers in direct ELISAs and Western blots. This antibody detects an epitope in the Lymphotoxin beta subunit. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Lymphotoxin alpha , rhAPRIL, rhBAFF, rhTNF-alpha , rhTWEAK, rhEDA-A2, rhEDA, rhTRAIL, rhVEGI, rhTRANCE, rhFas Ligand, or rhOX40 Ligand. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title