missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lymphotoxin beta/TNFSF3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1684-SP
This item is not returnable.
View return policy
Description
Lymphotoxin beta/TNFSF3 Monoclonal specifically detects Lymphotoxin beta/TNFSF3 in Human samples. It is validated for Western Blot.
Specifications
| Lymphotoxin beta/TNFSF3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| LTB, lymphotoxin beta (TNF superfamily, member 3), lymphotoxin-beta, p33, TNFCTNF-C, TNFSF3, TNFSF3LT-beta, Tumor necrosis factor C, Tumor necrosis factor ligand superfamily member 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4050 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 135105 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| LTB | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Lymphotoxin alpha/beta | |
| 25 μg | |
| Primary | |
| Detects human Lymphotoxin alpha beta heterotrimers in direct ELISAs and Western blots. This antibody detects an epitope in the Lymphotoxin beta subunit. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) Lymphotoxin alpha , rhAPRIL, rhBAFF, rhTNF-alpha , rhTWEAK, rhEDA-A2, rhEDA, rhTRAIL, rhVEGI, rhTRANCE, rhFas Ligand, or rhOX40 Ligand. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction