missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lipocalin-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
£157.00 - £425.00
Specifications
| Antigen | Lipocalin-1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18766257
![]() |
R&D Systems
AF1708-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18040952
|
R&D Systems
AF1708 |
100 μg |
£425.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lipocalin-1 Polyclonal specifically detects Lipocalin-1 in Human samples. It is validated for Western Blot.Specifications
| Lipocalin-1 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| LCN1, lipocalin 1 (tear prealbumin), lipocalin 1-like 2, lipocalin-1, Lipocalin1, MGC71975, PMFA, Tear lipocalin, Tear prealbumin, Tlc, TPVon Ebner gland protein, VEG protein, VEGP, VEGPlipocalin 1 (protein migrating faster than albumin, tear prealbumin) | |
| LCN1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P31025 | |
| 3933 | |
| Mouse myeloma cell line NS0-derived recombinant human Lipocalin-1 (R&D Systems, Catalog # 1708-PI) His19-Asp176 Accession # P31025 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Lipocalin-1 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human Lipocalin-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title