missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lipocalin-1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1708
This item is not returnable.
View return policy
Description
Lipocalin-1 Polyclonal specifically detects Lipocalin-1 in Human samples. It is validated for Western Blot.
Specifications
| Lipocalin-1 | |
| Polyclonal | |
| Unconjugated | |
| P31025 | |
| LCN1 | |
| Mouse myeloma cell line NS0-derived recombinant human Lipocalin-1 (R&D Systems, Catalog # 1708-PI) His19-Asp176 Accession # P31025 | |
| 100 μg | |
| Primary | |
| Detects human Lipocalin-1 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human Lipocalin-2 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| LCN1, lipocalin 1 (tear prealbumin), lipocalin 1-like 2, lipocalin-1, Lipocalin1, MGC71975, PMFA, Tear lipocalin, Tear prealbumin, Tlc, TPVon Ebner gland protein, VEG protein, VEGP, VEGPlipocalin 1 (protein migrating faster than albumin, tear prealbumin) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3933 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction