missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LIMPII/SR-B2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
£167.00 - £474.00
Specifications
| Antigen | LIMPII/SR-B2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18768977
![]() |
R&D Systems
AF1966-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18028531
|
R&D Systems
AF1966 |
100 μg |
£474.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIMPII/SR-B2 Polyclonal specifically detects LIMPII/SR-B2 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| LIMPII/SR-B2 | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2, CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, LPG85, lysosome membrane protein 2, Lysosome membrane protein II, SCARB2, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SRB2, SR-B2, SR-BII | |
| SCARB2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Q14108 | |
| 950 | |
| Mouse myeloma cell line NS0-derived recombinant human LIMPII lumenal loop Arg27-Thr432 Accession # Q14108 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human LIMPII in ELISAs and Western blots. In sandwich immunoassays, approximately 6% cross-reactivity with recombinant mouse LIMPII is observed and less than 0.3% cross-reactivity with recombinant human (rh) SR-B1 and rhCD36 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title