missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LIMPII/SR-B2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF1966-SP
This item is not returnable.
View return policy
Description
LIMPII/SR-B2 Polyclonal specifically detects LIMPII/SR-B2 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| LIMPII/SR-B2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2, CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, LPG85, lysosome membrane protein 2, Lysosome membrane protein II, SCARB2, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SRB2, SR-B2, SR-BII | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 950 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Q14108 | |
| SCARB2 | |
| Mouse myeloma cell line NS0-derived recombinant human LIMPII lumenal loop Arg27-Thr432 Accession # Q14108 | |
| 25 μg | |
| Primary | |
| Detects human LIMPII in ELISAs and Western blots. In sandwich immunoassays, approximately 6% cross-reactivity with recombinant mouse LIMPII is observed and less than 0.3% cross-reactivity with recombinant human (rh) SR-B1 and rhCD36 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction