missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB5/CD85c/LIR-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £369.00
Specifications
| Antigen | LIR-8/CD85c/LILRB5 |
|---|---|
| Clone | 395239 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18760397
![]() |
R&D Systems
MAB3065-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18170811
![]() |
R&D Systems
MAB3065 |
100 μg |
£369.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIR-8/CD85c/LILRB5 Monoclonal specifically detects LIR-8/CD85c/LILRB5 in Human samples. It is validated for Western Blot.Specifications
| LIR-8/CD85c/LILRB5 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O75023 | |
| 10990 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRB5/CD85c/LIR-8 isoform 1 Arg18-His456 Accession # O75023 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human LILRB5/CD85c/LIR-8 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ILT1, 2, 3, 4, 5, rhLIR-6, or rhILT11/LILRA5 is observed. In Western blots, less than 5% cross-reactivity with rhILT2, 3, 5, rhLIR-6 or rhILT11/LILRA5 is observed, and no cross-reactivity with rhILT1, 4, or 6. |
| 395239 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD85 antigen-like family member C, CD85c, CD85c antigen, Leukocyte immunoglobulin-like receptor 8, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR8, LIR-8, LIR8CD85C, LIR-8leukocyte immunoglobulin-like receptor subfamily B member 5, member 5 | |
| LILRB5 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title