missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRB5/CD85c/LIR-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3065-SP
This item is not returnable.
View return policy
Description
LIR-8/CD85c/LILRB5 Monoclonal specifically detects LIR-8/CD85c/LILRB5 in Human samples. It is validated for Western Blot.
Specifications
| LIR-8/CD85c/LILRB5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O75023 | |
| LILRB5 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRB5/CD85c/LIR-8 isoform 1 Arg18-His456 Accession # O75023 | |
| 25 μg | |
| Primary | |
| Detects human LILRB5/CD85c/LIR-8 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ILT1, 2, 3, 4, 5, rhLIR-6, or rhILT11/LILRA5 is observed. In Western blots, less than 5% cross-reactivity with rhILT2, 3, 5, rhLIR-6 or rhILT11/LILRA5 is observed, and no cross-reactivity with rhILT1, 4, or 6. | |
| Human | |
| Purified |
| Western Blot | |
| 395239 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD85 antigen-like family member C, CD85c, CD85c antigen, Leukocyte immunoglobulin-like receptor 8, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR8, LIR-8, LIR8CD85C, LIR-8leukocyte immunoglobulin-like receptor subfamily B member 5, member 5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10990 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction