missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Layilin Antibody, R&D Systems™

Mouse Monoclonal Antibody
£170.00 - £506.00
Specifications
| Antigen | Layilin |
|---|---|
| Clone | 328002 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18797519
![]() |
R&D Systems
MAB26371-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18129410
![]() |
R&D Systems
MAB26371 |
500 μg |
£506.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Layilin | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ30977, FLJ31092, layilin, LAYN, LYAN | |
| LAYN | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 328002 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_849156 | |
| 143903 | |
| Mouse myeloma cell line NS0-derived recombinant human Layilin Ala22-Glu220 Accession # NP_849156 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Layilin in ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title