missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Layilin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB26371-SP
This item is not returnable.
View return policy
Description
Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| Layilin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ30977, FLJ31092, layilin, LAYN, LYAN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 143903 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 328002 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_849156 | |
| LAYN | |
| Mouse myeloma cell line NS0-derived recombinant human Layilin Ala22-Glu220 Accession # NP_849156 | |
| 25 μg | |
| Primary | |
| Detects human Layilin in ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction