missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Latent TGF-beta bp1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
£180.00 - £1790.00
Specifications
| Antigen | Latent TGF-beta bp1 |
|---|---|
| Clone | 35409 |
| Dilution | Western Blot 1 ug/mL, Immunoaffinity Purification |
| Applications | Western Blot, Affinity Purification |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18771898
![]() |
R&D Systems
MAB388-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18780988
![]() |
R&D Systems
MAB388-100 |
100 μg |
£462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783359
![]() |
R&D Systems
MAB388-500 |
500 μg |
£1790.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Latent TGF-beta bp1 Monoclonal specifically detects Latent TGF-beta bp1 in Human samples. It is validated for Western Blot, Immunoaffinity Purification.Specifications
| Latent TGF-beta bp1 | |
| Western Blot 1 ug/mL, Immunoaffinity Purification | |
| Monoclonal | |
| Mouse | |
| Human | |
| Latent TGFbeta bp1, latent transforming growth factor beta binding protein 1, latent-transforming growth factor beta-binding protein 1, LTBP1, LTBP-1, MGC163161, TGF-beta1-BP-1, Transforming growth factor beta-1-binding protein 1 | |
| LTBP1 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 35409 | |
| Western Blot, Affinity Purification | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 4052 | |
| Human platelet-derived large latent TGF-beta 1 complex | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human LTBP-1 from the large latent TGF-beta 1 complex. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title