missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Latent TGF-beta bp1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems MAB388-100
This item is not returnable.
View return policy
Description
Latent TGF-beta bp1 Monoclonal specifically detects Latent TGF-beta bp1 in Human samples. It is validated for Western Blot, Immunoaffinity Purification.
Specifications
| Latent TGF-beta bp1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| LTBP1 | |
| Human platelet-derived large latent TGF-beta 1 complex | |
| 100 μg | |
| Primary | |
| Detects human LTBP-1 from the large latent TGF-beta 1 complex. | |
| Human | |
| Ascites |
| Western Blot, Affinity Purification | |
| 35409 | |
| Western Blot 1 ug/mL, Immunoaffinity Purification | |
| Latent TGFbeta bp1, latent transforming growth factor beta binding protein 1, latent-transforming growth factor beta-binding protein 1, LTBP1, LTBP-1, MGC163161, TGF-beta1-BP-1, Transforming growth factor beta-1-binding protein 1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 4052 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction