missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIRREL3 Full-length ORF (NP_115920.1, 1 a.a. - 778 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084623-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135983

  • £280.00 / 10µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the nephrin-like protein family. These proteins are expressed in fetal and adult brain, and also in podocytes of kidney glomeruli. The cytoplasmic domains of these proteins interact with the C-terminus of podocin, also expressed in the podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. Mutations in this gene are associated with mental retardation autosomal dominant type 4 (MRD4). Alternatively spliced transcript variants encoding different isoforms have been found for this gene

Sequence: MKPFQLDLLFVCFFLFSQELGLQKRGCCLVLGYMAKDKFRRMNEGQVYSFSQQPQDQVVVSGQPVTLLCAIPEYDGFVLWIKDGLALGVGRDLSSYPQYLVVGNHLSGEHHLKILRAELQDDAVYECQAIQAAIRSRPARLTVLVPPDDPVILGGPVISLRAGDPLNLTCHADNAKPAASIIWLRKGEVINGATYSKTLLRDGKRESIVSTLFISPGDVENGQSIVCRATNKAIPGGKETSVTIDIQHPPLVNLSVEPQPVLEDNVVTFHCSAKANPAVTQYRWAKRGQIIKEASGEVYRTTVDYTYFSEPVSCEVTNALGSTNLSRTVDVYFGPRMTTEPQSLLVDLGSDAIFSCAWTGNPSLTIVWMKRGSGVVLSNEKTLTLKSVRQEDAGKYVCRAVVPRVGAGEREVTLTVNGPPIISSTQTQHALHGEKGQIKCFIRSTPPPDRIAWSWKENVLESGTSGRYTVETISTEEGVISTLTISNIVRADFQTIYNCTAWNSFGSDTEIIRLKEQGSEMKSGAGLEAESVPMAVIIGVAVGAGVAFLVLMATIVAFCCARSQRNLKGVVSAKNDIRVEIVHKEPASGREGEEHSTIKQLMMDRGEFQQDSVLKQLEVLKEEEKEFQNLKDPTNGYYSVNTFKEHHSTPTISLSSCQPDLRPAGKQRVPTGMSFTNIYSTLSGQGRLYDYGQRFVLGMGSSSIELCEREFQRGSLSDSSSFLDTQCDSSVSSSGKQDGYVQFDKASKASASSSHHSQSSSQNSDPSRPLQRRMQTHV
Specifications

Specifications

NP_115920.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
111.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1867/KIRRE/MGC126824/MGC126850/NEPH2/PRO4502
KIRREL3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84623
KIRREL3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKPFQLDLLFVCFFLFSQELGLQKRGCCLVLGYMAKDKFRRMNEGQVYSFSQQPQDQVVVSGQPVTLLCAIPEYDGFVLWIKDGLALGVGRDLSSYPQYLVVGNHLSGEHHLKILRAELQDDAVYECQAIQAAIRSRPARLTVLVPPDDPVILGGPVISLRAGDPLNLTCHADNAKPAASIIWLRKGEVINGATYSKTLLRDGKRESIVSTLFISPGDVENGQSIVCRATNKAIPGGKETSVTIDIQHPPLVNLSVEPQPVLEDNVVTFHCSAKANPAVTQYRWAKRGQIIKEASGEVYRTTVDYTYFSEPVSCEVTNALGSTNLSRTVDVYFGPRMTTEPQSLLVDLGSDAIFSCAWTGNPSLTIVWMKRGSGVVLSNEKTLTLKSVRQEDAGKYVCRAVVPRVGAGEREVTLTVNGPPIISSTQTQHALHGEKGQIKCFIRSTPPPDRIAWSWKENVLESGTSGRYTVETISTEEGVISTLTISNIVRADFQTIYNCTAWNSFGSDTEIIRLKEQGSEMKSGAGLEAESVPMAVIIGVAVGAGVAFLVLMATIVAFCCARSQRNLKGVVSAKNDIRVEIVHKEPASGREGEEHSTIKQLMMDRGEFQQDSVLKQLEVLKEEEKEFQNLKDPTNGYYSVNTFKEHHSTPTISLSSCQPDLRPAGKQRVPTGMSFTNIYSTLSGQGRLYDYGQRFVLGMGSSSIELCEREFQRGSLSDSSSFLDTQCDSSVSSSGKQDGYVQFDKASKASASSSHHSQSSSQNSDPSRPLQRRMQTHV
RUO
KIRREL3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers