Learn More
Abnova™ Human KIRREL3 Full-length ORF (NP_115920.1, 1 a.a. - 778 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
£280.00 - £425.00
Specifications
Accession Number | NP_115920.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84623 |
Molecular Weight (g/mol) | 111.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16145983
|
Abnova™
H00084623-P01.25UG |
25 ug |
£425.00
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16135983
|
Abnova™
H00084623-P01.10UG |
10 ug |
£280.00
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the nephrin-like protein family. These proteins are expressed in fetal and adult brain, and also in podocytes of kidney glomeruli. The cytoplasmic domains of these proteins interact with the C-terminus of podocin, also expressed in the podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. Mutations in this gene are associated with mental retardation autosomal dominant type 4 (MRD4). Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Sequence: MKPFQLDLLFVCFFLFSQELGLQKRGCCLVLGYMAKDKFRRMNEGQVYSFSQQPQDQVVVSGQPVTLLCAIPEYDGFVLWIKDGLALGVGRDLSSYPQYLVVGNHLSGEHHLKILRAELQDDAVYECQAIQAAIRSRPARLTVLVPPDDPVILGGPVISLRAGDPLNLTCHADNAKPAASIIWLRKGEVINGATYSKTLLRDGKRESIVSTLFISPGDVENGQSIVCRATNKAIPGGKETSVTIDIQHPPLVNLSVEPQPVLEDNVVTFHCSAKANPAVTQYRWAKRGQIIKEASGEVYRTTVDYTYFSEPVSCEVTNALGSTNLSRTVDVYFGPRMTTEPQSLLVDLGSDAIFSCAWTGNPSLTIVWMKRGSGVVLSNEKTLTLKSVRQEDAGKYVCRAVVPRVGAGEREVTLTVNGPPIISSTQTQHALHGEKGQIKCFIRSTPPPDRIAWSWKENVLESGTSGRYTVETISTEEGVISTLTISNIVRADFQTIYNCTAWNSFGSDTEIIRLKEQGSEMKSGAGLEAESVPMAVIIGVAVGAGVAFLVLMATIVAFCCARSQRNLKGVVSAKNDIRVEIVHKEPASGREGEEHSTIKQLMMDRGEFQQDSVLKQLEVLKEEEKEFQNLKDPTNGYYSVNTFKEHHSTPTISLSSCQPDLRPAGKQRVPTGMSFTNIYSTLSGQGRLYDYGQRFVLGMGSSSIELCEREFQRGSLSDSSSFLDTQCDSSVSSSGKQDGYVQFDKASKASASSSHHSQSSSQNSDPSRPLQRRMQTHVSpecifications
NP_115920.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
111.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKPFQLDLLFVCFFLFSQELGLQKRGCCLVLGYMAKDKFRRMNEGQVYSFSQQPQDQVVVSGQPVTLLCAIPEYDGFVLWIKDGLALGVGRDLSSYPQYLVVGNHLSGEHHLKILRAELQDDAVYECQAIQAAIRSRPARLTVLVPPDDPVILGGPVISLRAGDPLNLTCHADNAKPAASIIWLRKGEVINGATYSKTLLRDGKRESIVSTLFISPGDVENGQSIVCRATNKAIPGGKETSVTIDIQHPPLVNLSVEPQPVLEDNVVTFHCSAKANPAVTQYRWAKRGQIIKEASGEVYRTTVDYTYFSEPVSCEVTNALGSTNLSRTVDVYFGPRMTTEPQSLLVDLGSDAIFSCAWTGNPSLTIVWMKRGSGVVLSNEKTLTLKSVRQEDAGKYVCRAVVPRVGAGEREVTLTVNGPPIISSTQTQHALHGEKGQIKCFIRSTPPPDRIAWSWKENVLESGTSGRYTVETISTEEGVISTLTISNIVRADFQTIYNCTAWNSFGSDTEIIRLKEQGSEMKSGAGLEAESVPMAVIIGVAVGAGVAFLVLMATIVAFCCARSQRNLKGVVSAKNDIRVEIVHKEPASGREGEEHSTIKQLMMDRGEFQQDSVLKQLEVLKEEEKEFQNLKDPTNGYYSVNTFKEHHSTPTISLSSCQPDLRPAGKQRVPTGMSFTNIYSTLSGQGRLYDYGQRFVLGMGSSSIELCEREFQRGSLSDSSSFLDTQCDSSVSSSGKQDGYVQFDKASKASASSSHHSQSSSQNSDPSRPLQRRMQTHV | |
RUO | |
KIRREL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84623 | |
KIRREL3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1867/KIRRE/MGC126824/MGC126850/NEPH2/PRO4502 | |
KIRREL3 | |
Recombinant | |
wheat germ expression system |