missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DS5/CD158g Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£178.00 - £362.00
Specifications
| Antigen | KIR2DS5/CD158g |
|---|---|
| Clone | 1165A |
| Dilution | Flow Cytometry 1:1000 dilution |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18791948
![]() |
R&D Systems
MAB8920-SP |
25 μg |
£178.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18142791
|
R&D Systems
MAB8920 |
100 μg |
£362.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIR2DS5/CD158g Monoclonal specifically detects KIR2DS5/CD158g in Human samples. It is validated for Flow Cytometry.Specifications
| KIR2DS5/CD158g | |
| Flow Cytometry 1:1000 dilution | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| killer cell immunoglobulin-like receptor 2DS5, CD158g, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5, NKAT9 | |
| KIR2DS5 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Detects human KIR2DS5/CD158g in direct ELISAs. |
| 1165A | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q14953 | |
| 3810 | |
| NS0 mouse myeloma cell line transfected with human KIR2DS5/CD158g His22-His245 Accession # Q14953 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title