missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DS5/CD158g Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8920-SP
This item is not returnable.
View return policy
Description
KIR2DS5/CD158g Monoclonal specifically detects KIR2DS5/CD158g in Human samples. It is validated for Flow Cytometry.
Specifications
| KIR2DS5/CD158g | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| killer cell immunoglobulin-like receptor 2DS5, CD158g, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5, NKAT9 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3810 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1165A | |
| Flow Cytometry 1:1000 dilution | |
| Q14953 | |
| KIR2DS5 | |
| NS0 mouse myeloma cell line transfected with human KIR2DS5/CD158g His22-His245 Accession # Q14953 | |
| 25 μg | |
| Primary | |
| Detects human KIR2DS5/CD158g in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction