missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KGF/FGF-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£170.00 - £1765.00
Specifications
| Antigen | KGF/FGF-7 |
|---|---|
| Clone | 29522 |
| Dilution | Neutralization 0.1-0.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746489
![]() |
R&D Systems
MAB251-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732568
![]() |
R&D Systems
MAB251-100 |
100 μg |
£446.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18776409
![]() |
R&D Systems
MAB251-500 |
500 μg |
£1765.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KGF/FGF-7 Monoclonal specifically detects KGF/FGF-7 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| KGF/FGF-7 | |
| Neutralization 0.1-0.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) | |
| FGF7 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 29522 | |
| Neutralization, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P21781 | |
| 2252 | |
| E. coli-derived recombinant human KGF/FGF-7 Cys32-Thr194 Accession # P21781 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human KGF/FGF-7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) FGF-3, rhFGF-4, rhFGF-5, rhFGF-6, rhFGF acidic, or rhFGF basic is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title