missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KGF/FGF-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB251-500
This item is not returnable.
View return policy
Description
KGF/FGF-7 Monoclonal specifically detects KGF/FGF-7 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| KGF/FGF-7 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2252 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization, ELISA | |
| 29522 | |
| Neutralization 0.1-0.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P21781 | |
| FGF7 | |
| E. coli-derived recombinant human KGF/FGF-7 Cys32-Thr194 Accession # P21781 | |
| 500 μg | |
| Primary | |
| Detects human KGF/FGF-7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) FGF-3, rhFGF-4, rhFGF-5, rhFGF-6, rhFGF acidic, or rhFGF basic is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction