missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 8/Neuropsin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
£156.00 - £460.00
Specifications
| Antigen | Kallikrein 8/Neuropsin |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Neutralization 8 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18748937
![]() |
R&D Systems
AF2025-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18003422
|
R&D Systems
AF2025 |
100 μg |
£460.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kallikrein 8/Neuropsin Polyclonal specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for Western Blot, Neutralization.Specifications
| Kallikrein 8/Neuropsin | |
| Western Blot 0.1 ug/mL, Neutralization 8 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein | |
| KLK8 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| O60259 | |
| 11202 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 8 Gln29-Gly260 Accession # O60259 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Kallikrein 8/Neuropsin in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title