missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 8/Neuropsin Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2025
This item is not returnable.
View return policy
Description
Kallikrein 8/Neuropsin Polyclonal specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| Kallikrein 8/Neuropsin | |
| Polyclonal | |
| Unconjugated | |
| O60259 | |
| KLK8 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 8 Gln29-Gly260 Accession # O60259 | |
| 100 μg | |
| Primary | |
| Detects human Kallikrein 8/Neuropsin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 8 ug/mL | |
| EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11202 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction