missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB4104
This item is not returnable.
View return policy
Description
Kallikrein 2 Monoclonal specifically detects Kallikrein 2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.
Specifications
| Kallikrein 2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 3.4 ug/mL | |
| P20151 | |
| KLK2 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 2 Ile25-Pro261 Accession # P20151 | |
| 100 μg | |
| Primary | |
| Detects human Kallikrein 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Kallikrein 1, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, 15, or B1 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Neutralization | |
| 426723 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, Klk1c2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2, Ton | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3817 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction