missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £398.00
Specifications
| Antigen | Kallikrein 2 |
|---|---|
| Clone | 426723 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 3.4 ug/mL |
| Applications | Western Blot, Immunoprecipitation, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18723909
![]() |
R&D Systems
MAB4104-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109900
|
R&D Systems
MAB4104 |
100 μg |
£398.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kallikrein 2 Monoclonal specifically detects Kallikrein 2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
| Kallikrein 2 | |
| LYOPH | |
| Western Blot, Immunoprecipitation, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| P20151 | |
| 3817 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 2 Ile25-Pro261 Accession # P20151 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Kallikrein 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Kallikrein 1, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, 15, or B1 is observed. |
| 426723 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 3.4 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, Klk1c2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2, Ton | |
| KLK2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title