missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 15 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £478.00
Specifications
| Antigen | Kallikrein 15 |
|---|---|
| Clone | 252819 |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18755468
![]() |
R&D Systems
MAB2540-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18180811
|
R&D Systems
MAB2540 |
500 μg |
£478.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kallikrein 15 Monoclonal specifically detects Kallikrein 15 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Kallikrein 15 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 55554 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 15 isoform 1 Leu23-Asn256 (predicted) | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Kallikrein 15 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) Factor X is observed and no cross-reactivity with recombinant mouse HGFA, rhKallikrein 3, 5, 6, 8, 10, 11, rhThrombin, or rhuPA is observed. |
| 252819 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| ACO, ACO protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, HSRNASPH, kallikrein 15, kallikrein-15, kallikrein-like serine protease, kallikrein-related peptidase 15, KLK15, Prostinogen | |
| KLK15 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title