missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 15 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2540
This item is not returnable.
View return policy
Description
Kallikrein 15 Monoclonal specifically detects Kallikrein 15 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Kallikrein 15 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KLK15 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 15 isoform 1 Leu23-Asn256 (predicted) | |
| 500 μg | |
| Primary | |
| Detects human Kallikrein 15 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) Factor X is observed and no cross-reactivity with recombinant mouse HGFA, rhKallikrein 3, 5, 6, 8, 10, 11, rhThrombin, or rhuPA is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 252819 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| ACO, ACO protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, HSRNASPH, kallikrein 15, kallikrein-15, kallikrein-like serine protease, kallikrein-related peptidase 15, KLK15, Prostinogen | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55554 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction