missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Jagged 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £1805.00
Specifications
| Antigen | Jagged 1 |
|---|---|
| Clone | 188323 |
| Concentration | LYOPH |
| Dilution | Neutralization 0.5-3 ug/mL |
| Applications | Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773339
![]() |
R&D Systems
MAB12771-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18791939
![]() |
R&D Systems
MAB12771-100 |
100 μg |
£449.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18725078
![]() |
R&D Systems
MAB12771-500 |
500 μg |
£1805.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Jagged 1 Monoclonal specifically detects Jagged 1 in Human samples. It is validated for Neutralization.Specifications
| Jagged 1 | |
| LYOPH | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| P78504 | |
| 182 | |
| NS0-derived recombinant human Jagged 1 Ser32-Asp296 Accession # P78504 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Jagged 1 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human Jagged 2 and recombinant rat Jagged 1 is observed. |
| 188323 | |
| Neutralization 0.5-3 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, JAG1, jagged 1, Jagged1, JAGL1, protein jagged-1 | |
| JAG1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title