missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Jagged 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB12771-500
This item is not returnable.
View return policy
Description
Jagged 1 Monoclonal specifically detects Jagged 1 in Human samples. It is validated for Neutralization.
Specifications
| Jagged 1 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.5-3 ug/mL | |
| P78504 | |
| JAG1 | |
| NS0-derived recombinant human Jagged 1 Ser32-Asp296 Accession # P78504 | |
| 500 μg | |
| Primary | |
| Detects human Jagged 1 in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant human Jagged 2 and recombinant rat Jagged 1 is observed. | |
| Human | |
| Purified |
| Neutralization | |
| 188323 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, JAG1, jagged 1, Jagged1, JAGL1, protein jagged-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 182 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction