missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Isocitrate Dehydrogenase 2/IDH2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£195.00 - £460.00
Specifications
| Antigen | Isocitrate Dehydrogenase 2/IDH2 |
|---|---|
| Clone | 1069032 |
| Dilution | Immunohistochemistry 3-25 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, Immunocytochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18754414
|
R&D Systems
MAB11462-100 |
100 μg |
£460.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18704344
|
R&D Systems
MAB11462-SP |
25 μg |
£195.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isocitrate Dehydrogenase 2/IDH2 Monoclonal antibody specifically detects Isocitrate Dehydrogenase 2/IDH2 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), ImmunocytochemistrySpecifications
| Isocitrate Dehydrogenase 2/IDH2 | |
| Immunohistochemistry 3-25 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 3418 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1069032 | |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P48735 | |
| E. coli -derived recombinant human IDH-2, Thr376-Gln452, Accession # P48735 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title