missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Isocitrate Dehydrogenase 2/IDH2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11462-100
This item is not returnable.
View return policy
Description
Isocitrate Dehydrogenase 2/IDH2 Monoclonal antibody specifically detects Isocitrate Dehydrogenase 2/IDH2 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Specifications
| Isocitrate Dehydrogenase 2/IDH2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3418 | |
| Human | |
| Purified |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 1069032 | |
| Immunohistochemistry 3-25 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, Immunocytochemistry 3-25 μg/mL | |
| P48735 | |
| E. coli -derived recombinant human IDH-2, Thr376-Gln452, Accession # P48735 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction