missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin beta 6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £362.00
Specifications
| Antigen | Integrin beta 6 |
|---|---|
| Clone | 437216 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18705409
![]() |
R&D Systems
MAB41551-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18152221
![]() |
R&D Systems
MAB41551 |
100 μg |
£362.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin beta 6 Monoclonal specifically detects Integrin beta 6 in Human samples. It is validated for Western Blot.Specifications
| Integrin beta 6 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P18564 | |
| 3694 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta 6 Gly22-Asn707 Accession # P18564 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Integrin beta 6 in direct ELISAs and Western blots. In Western blots (under reducing conditions), 40% cross-reactivity with recombinant human (rh) Integrin beta 2 is observed and no cross-reactivity with rhIntegrin beta 1, 3, 5, 7, or 8 is observed. |
| 437216 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| integrin beta-6, integrin, beta 6, ITGB6 | |
| ITGB6 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title