missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin beta 6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB41551
This item is not returnable.
View return policy
Description
Integrin beta 6 Monoclonal specifically detects Integrin beta 6 in Human samples. It is validated for Western Blot.
Specifications
| Integrin beta 6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P18564 | |
| ITGB6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta 6 Gly22-Asn707 Accession # P18564 | |
| 100 μg | |
| Primary | |
| Detects human Integrin beta 6 in direct ELISAs and Western blots. In Western blots (under reducing conditions), 40% cross-reactivity with recombinant human (rh) Integrin beta 2 is observed and no cross-reactivity with rhIntegrin beta 1, 3, 5, 7, or 8 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 437216 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| integrin beta-6, integrin, beta 6, ITGB6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3694 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction