missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-6R alpha (Research Grade Tocilizumab Biosimilar) Antibody, R&D Systems™
Human Monoclonal Antibody
£218.00 - £500.00
Specifications
| Antigen | IL-6R alpha |
|---|---|
| Clone | Hu203 |
| Dilution | Flow Cytometry 0.25 μg/10^6 cells, ELISA, Blockade of Receptor-ligand Interaction 0.25 μg/10^6 cells |
| Applications | Flow Cytometry, ELISA, Blockade of Receptor-ligand Interaction |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18794404
|
R&D Systems
MAB11482-SP |
25 μg |
£218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18754304
|
R&D Systems
MAB11482-100 |
100 μg |
£500.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry, ELISA, Blockade of Receptor-ligand InteractionSpecifications
| IL-6R alpha | |
| Flow Cytometry 0.25 μg/10^6 cells, ELISA, Blockade of Receptor-ligand Interaction 0.25 μg/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
| E. coli-derived recombinant human IL-6R alpha, Arg387-Arg468, Accession # P08887 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Hu203 | |
| Flow Cytometry, ELISA, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| Human | |
| Human | |
| P08887 | |
| 3570 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title