Learn More
Human IL-6R alpha (Research Grade Tocilizumab Biosimilar) Antibody, R&D Systems™
Human Monoclonal Antibody
Brand:  R&D Systems MAB11482-100
 View more versions of this product
Product Code. 18754304
  • £500.00 / 100µg
Description
IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry, ELISA, Blockade of Receptor-ligand Interaction
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-6R alpha
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
E. coli-derived recombinant human IL-6R alpha, Arg387-Arg468, Accession # P08887
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Flow Cytometry, ELISA, Blockade of Receptor-ligand Interaction
Hu203
Flow Cytometry 0.25 μg/10^6 cells, ELISA, Blockade of Receptor-ligand Interaction 0.25 μg/10^6 cells
P08887
Human
Protein A or G purified from hybridoma culture supernatant
RUO
3570
Human
Purified