missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-6 Recombinant Protein
Recombinant Human Interleukin 6 (IL6) is a bioactive protein intended for use in cell culture applications
£72.25 - £2680.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 3569 |
Molecular Weight (g/mol) | 21.3 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | IL-6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10242823
|
Gibco™
PHC0064 |
5μg |
£72.25
Each |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10079162
|
Gibco™
PHC0065 |
10μg |
£101.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10089162
|
Gibco™
PHC0066 |
25μg |
£194.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10777823
|
Gibco™
PHC0061 |
100μg |
£656.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10690144
|
Gibco™
PHC0063 |
1mg |
£2680.00
Each |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Human IL-6 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL6 is a pleiotropic cytokine involved in several cellular processes including inflammation, hematopoeisis, and immune response.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse B9 cells
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
21.3 kDa | |
IL-6 | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Stem Cell Research, Inflammation, Bone Research | |
95 % |
3569 | |
Sequential Chromatography | |
Human | |
Carrier-Free | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.