missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-6 Recombinant Protein

Recombinant Human Interleukin 6 (IL6) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC0063

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10690144

  • £2680.00 / Each
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Human IL-6 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IL6 is a pleiotropic cytokine involved in several cellular processes including inflammation, hematopoeisis, and immune response.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse B9 cells
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture
21.3 kDa
1mg
Human
Carrier-Free
1 mg
E. coli
Recombinant, Ligand
Interleukins
Gibco™
3569
Sequential Chromatography
IL-6
Store in refrigerator (2–8°C).
< 0.1 ng/μg
Wet Ice
Cytokines & Receptors
Lyophilized
Immunology, Stem Cell Research, Inflammation, Bone Research
95 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.