missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-15 Quantikine ELISA Kit

Product Code. 18762304
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Sufficient For unitSize
18762304 1 Plate 1 x 96-well Plate 1 set
18761854 6 Plates 6 x 96-well Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18762304 Supplier R&D Systems™ Supplier No. D1500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human IL-15 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IL-15 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-15 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.90 - 250pg/mL
Assay Sensitivity 2pg/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Human IL-15
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3600
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym IL15, IL-15 mgC9721, interleukin 15, interleukin-15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.