missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-11 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£167.00 - £1775.00
Specifications
| Antigen | IL-11 |
|---|---|
| Clone | 22616 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731289
![]() |
R&D Systems
MAB618-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702538
![]() |
R&D Systems
MAB618-100 |
100 μg |
£448.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18791919
![]() |
R&D Systems
MAB618-500 |
500 μg |
£1775.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-11 Monoclonal specifically detects IL-11 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| IL-11 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P20809 | |
| 3589 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-11 Pro22-Leu199 Accession # P20809 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-11 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse IL-11 but does not cross-react with recombinant human (rh) IL-6, rhIL-6 R, rhLIF, rhOSM, rhCT-1, rhCNTF, or rhCLC. |
| 22616 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin | |
| IL11 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title