missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-11 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB618-500
This item is not returnable.
View return policy
Description
IL-11 Monoclonal specifically detects IL-11 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-11 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3589 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, ELISA | |
| 22616 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P20809 | |
| IL11 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-11 Pro22-Leu199 Accession # P20809 | |
| 500 μg | |
| Primary | |
| Detects human IL-11 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse IL-11 but does not cross-react with recombinant human (rh) IL-6, rhIL-6 R, rhLIF, rhOSM, rhCT-1, rhCNTF, or rhCLC. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction