missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-10 Recombinant Protein
Recombinant IL10 is a bioactive protein intended for use in cell culture applications
£115.00 - £4260.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 18.6 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | IL-10 |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10180763
|
Gibco™
PHC0104 |
5μg |
£115.00
Each |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10680534
|
Gibco™
PHC0105 |
10μg |
£195.00
Each |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10476093
|
Gibco™
PHC0101 |
100μg |
£1528.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10486093
|
Gibco™
PHC0103 |
1mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Human IL-10 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL10 inhibits the synthesis of a number of cytokines, such as TNF-α, IL-1, and IL-6 from macrophages and IL-2, IFN-γ, and TNF-β from Th1 cells.Approved for shipment on Wet or Dry Ice.
Specifications
Cell Culture | |
Sequential Chromatography | |
Human | |
Carrier-Free | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Signal Transduction, Stem Cell Research, Inflammation, Angiogenesis | |
95 % |
18.6 kDa | |
IL-10 | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.