missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-10 R beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £467.00
Specifications
| Antigen | IL-10 R beta |
|---|---|
| Clone | 90227 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736318
![]() |
R&D Systems
MAB8741-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18149570
![]() |
R&D Systems
MAB8741 |
500 μg |
£467.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-10 R beta Monoclonal specifically detects IL-10 R beta in Human samples. It is validated for Western Blot.Specifications
| IL-10 R beta | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDw210b, CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL10R beta, IL-10R2, IL-10R2IL-10R subunit beta, IL10RB, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4 | |
| IL10RB | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 90227 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q08334 | |
| 3588 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-10 R beta Met20-Ser220 Accession # Q08334 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-10 R beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, ∼10% cross-reactivity with recombinant human (rh) IL-10 R and rhIFN-alpha/beta R1 is observed and no cross-reactivity with rhIFN-gamma RI and rhIFN-gamma R2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title