missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-10 R beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8741
This item is not returnable.
View return policy
Description
IL-10 R beta Monoclonal specifically detects IL-10 R beta in Human samples. It is validated for Western Blot.
Specifications
| IL-10 R beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDw210b, CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL10R beta, IL-10R2, IL-10R2IL-10R subunit beta, IL10RB, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3588 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 90227 | |
| Western Blot 1 ug/mL | |
| Q08334 | |
| IL10RB | |
| Mouse myeloma cell line NS0-derived recombinant human IL-10 R beta Met20-Ser220 Accession # Q08334 | |
| 500 μg | |
| Primary | |
| Detects human IL-10 R beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, ∼10% cross-reactivity with recombinant human (rh) IL-10 R and rhIFN-alpha/beta R1 is observed and no cross-reactivity with rhIFN-gamma RI and rhIFN-gamma R2 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction