missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-1 beta/IL-1F2 DuoSet ELISA
Description
The Human IL-1 beta/IL-1F2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-1 beta/IL-1F2.
Specifications
Specifications
| Product Type | ELISA Development Kit |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 3.91 to 250 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3553 |
| Synonym | catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta |
| Packaging Quantity | 5 Plates |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?