missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGFBP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
£168.00 - £1525.00
Specifications
| Antigen | IGFBP-1 |
|---|---|
| Clone | 33627 |
| Dilution | Western Blot 1 ug/mL, Neutralization 10-40 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18700738
![]() |
R&D Systems
MAB675-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18798868
![]() |
R&D Systems
MAB675-100 |
100 μg |
£387.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18712049
![]() |
R&D Systems
MAB675-500 |
500 μg |
£1525.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGFBP-1 Monoclonal specifically detects IGFBP-1 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| IGFBP-1 | |
| Western Blot 1 ug/mL, Neutralization 10-40 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP | |
| IGFBP1 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 33627 | |
| Western Blot, Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3484 | |
| E. coli-derived recombinant human IGFBP-1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IGFBP-1 in ELISAs and Western blots. In Western blots, detection of recombinant mouse IGFBP-1 is observed but no cross-reactivity with recombinant human (rh) IGFBP-2, rhIGFBP-3, rhIGFBP-4 or rhIGFBP-5 is observered. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title