missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGFBP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB675-SP
This item is not returnable.
View return policy
Description
IGFBP-1 Monoclonal specifically detects IGFBP-1 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| IGFBP-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IGFBP1 | |
| E. coli-derived recombinant human IGFBP-1 | |
| 25 μg | |
| Primary | |
| Detects human IGFBP-1 in ELISAs and Western blots. In Western blots, detection of recombinant mouse IGFBP-1 is observed but no cross-reactivity with recombinant human (rh) IGFBP-2, rhIGFBP-3, rhIGFBP-4 or rhIGFBP-5 is observered. | |
| Human | |
| Purified |
| Western Blot, Neutralization, ELISA | |
| 33627 | |
| Western Blot 1 ug/mL, Neutralization 10-40 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3484 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction