missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGF-II/IGF2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £1535.00
Specifications
| Antigen | IGF-II |
|---|---|
| Clone | 75014 |
| Dilution | Western Blot 2 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773097
![]() |
R&D Systems
MAB2921-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772477
![]() |
R&D Systems
MAB2921-100 |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18750587
![]() |
R&D Systems
MAB2921-500 |
500 μg |
£1535.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Human samples. It is validated for Western Blot.Specifications
| IGF-II | |
| Western Blot 2 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01344.1 | |
| 3481 | |
| E. coli-derived recombinant human IGF-II Ala25-Glu91 Accession # P01344.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IGF-II in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 10 - 50% cross-reactivity with recombinant mouse IGF-II. |
| 75014 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A | |
| IGF2 | |
| IgG2a | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title