missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGF-II/IGF2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2921-100
This item is not returnable.
View return policy
Description
IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Human samples. It is validated for Western Blot.
Specifications
| IGF-II | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3481 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 75014 | |
| Western Blot 2 ug/mL | |
| P01344.1 | |
| IGF2 | |
| E. coli-derived recombinant human IGF-II Ala25-Glu91 Accession # P01344.1 | |
| 100 μg | |
| Primary | |
| Detects human IGF-II in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 10 - 50% cross-reactivity with recombinant mouse IGF-II. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction