Learn More
Thermo Scientific™ Pierce™ Human IFN alpha ELISA Kits (Multi-Subtype)
Description
Assay IFN-a (14 out of 15 identified subtypes of human interferon alpha) levels in human cell culture media with these VeriKine Human IFN alpha ELISA Kits (Multi-Subtype).
Target: 14 of 15 subtypes of human interferon alpha (IFN-alpha): IFN-αA, IFN-α2, IFN-αD, IFN-αB2, IFN-αC, IFN-αG, IFN-αH, IFN-αI, IFN-αJ1, IFN-αK, IFN-α1, IFN-α4a, IFN-α4b, and IFN-αWA
Format: Colorimetric ELISA (TMB substrate) in clear 96-well plate
Measurement: Absorbance at 450nm
Assay Range: 12.5 to 500pg/mL (high sensitivity); 156 to 5000pg/mL (extended range)
Sensitivity: Undetermined
Precision: Intra-assay CV ≤ 8%; inter-assay CV ≤ 8%
Sample Types: Cell culture supernatant, tissue culture media
Total Assay Time: 3.25 hours
Sample Size: 100μL
Specificity: No significant cross-reaction with human IFN-β, IFN-γ or IFN-ω; mouse or rat IFN-α, IFN-β, or IFN-γ; bovine IFN-τ
Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, substrates, and stop solution.
Specifications
Specifications
| Assay Range | High Sensitivity: 12.5 to 500pg/mL; Extended Range: 156 to 5000pg/mL |
| Assay Sensitivity | Undetermined |
| Description | Human IFN alpha ELISA Kit (Multi-Subtype) |
| Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
| Sample Type | Cell Culture Supernatant, Tissue Culture Media |
| Specificity | No significant cross-reaction with human IFN-ß, IFN-g or IFN-w; mouse or rat IFN-a, IFN-ß, or IFN-g; bovine IFN-t |
| Sufficient For | 5 x 96 microplate wells |
| Kit Contents | Antibody Concentrate, 1 vial, Dilution Buffer, 250mL, HRP Conjugate Concentrate, 1 vial, Wash Solution Concentrate, 250mL, TMB Substrate Solution, 60mL, Stop Solution, 60mL, Plate Sealers, 20 ea, HRP Conjugate Diluent, 60mL, Precoated 96-well Plates, 5 ea, Human IFN Alpha Standard 10,000pg/mL, 2.5mL |
| Measurement | Absorbance at 450nm |
| Precision | Intra-assay CV ≤ 8%; inter-assay CV ≤ 8% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.