missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HLA-DR PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
£262.00 - £471.00
Specifications
| Antigen | HLA-DR |
|---|---|
| Clone | L203 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751679
![]() |
R&D Systems
FAB4869P-025 |
25 Tests |
£262.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18789706
![]() |
R&D Systems
FAB4869P-100 |
100 Tests |
£471.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
HLA-DR Monoclonal specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry.Spezifikation
| HLA-DR | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLADR, HLA-DRA, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MHC Class II DR, MLRW | |
| HLA-DRA | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human HLA-DR. |
| L203 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| P01903 | |
| 3122 | |
| RPMI 8866 human lymphoblastoid cells Accession # P01903 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts