missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HLA-DR PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems FAB4869P-100
This item is not returnable.
View return policy
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human samples. It is validated for Flow Cytometry.
Specifications
| HLA-DR | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLADR, HLA-DRA, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MHC Class II DR, MLRW | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3122 | |
| Human | |
| Purified |
| Flow Cytometry | |
| L203 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P01903 | |
| HLA-DRA | |
| RPMI 8866 human lymphoblastoid cells Accession # P01903 | |
| 100 Tests | |
| Primary | |
| Detects human HLA-DR. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction