missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HIN-1/SCGB3A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £370.00
Specifications
| Antigen | HIN-1/SCGB3A1 |
|---|---|
| Clone | 410219 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728099
![]() |
R&D Systems
MAB27901-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18111501
|
R&D Systems
MAB27901 |
100 μg |
£370.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIN-1/SCGB3A1 Monoclonal specifically detects HIN-1/SCGB3A1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| HIN-1/SCGB3A1 | |
| LYOPH | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96QR1 | |
| 92304 | |
| E. coli-derived recombinant human HIN-1/SCGB3A1 Phe21-Gly104 Accession # Q96QR1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human HIN-1/SCGB3A1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse HIN-1 is observed. |
| 410219 | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cytokine high in normal-1, Cytokine HIN-1, High in normal 1, HIN1, HIN-1, HIN1Pneumo secretory protein 2, LU105, PnSP-2, PNSP2, PnSP-2Uteroglobin-related protein 2, SCGB3A1, secretoglobin family 3A member 1, secretoglobin, family 3A, member 1, UGRP2, UGRP2MGC87867 | |
| SCGB3A1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title