missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HIN-1/SCGB3A1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB27901
This item is not returnable.
View return policy
Description
HIN-1/SCGB3A1 Monoclonal specifically detects HIN-1/SCGB3A1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| HIN-1/SCGB3A1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q96QR1 | |
| SCGB3A1 | |
| E. coli-derived recombinant human HIN-1/SCGB3A1 Phe21-Gly104 Accession # Q96QR1 | |
| 100 μg | |
| Primary | |
| Detects human HIN-1/SCGB3A1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse HIN-1 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 410219 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cytokine high in normal-1, Cytokine HIN-1, High in normal 1, HIN1, HIN-1, HIN1Pneumo secretory protein 2, LU105, PnSP-2, PNSP2, PnSP-2Uteroglobin-related protein 2, SCGB3A1, secretoglobin family 3A member 1, secretoglobin, family 3A, member 1, UGRP2, UGRP2MGC87867 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 92304 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction